4-Fluor-2-iodtoluol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 1-Fluor-3-iod-4-methylbenzol, 4-Fluor-2-iod-1-methylbenzol

006249-5G 006249-25G
FLUO006249-5GEA 36.9 EUR
FLUO006249-5G FLUO006249-25G
4-Fluor-2-iodtoluol
4-Fluor-2-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236.03 g/mol
MDL: MFCD00013709
CAS-Nummer: 13194-67-7
EINECS: 236-153-7
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR