2-Fluor-6-methoxybenzonitril

Lieferant: FLUOROCHEM

CFF623.5 006266-25G 006266-100G
CHMPCFF623.5EA 532 EUR
CHMPCFF623.5 FLUO006266-25G FLUO006266-100G
2-Fluor-6-methoxybenzonitril
2-Fluor-6-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
Siedepunkt: 245
Schmelzpunkt: 93…95 °C
MDL: MFCD00042291
CAS-Nummer: 94088-46-7
EINECS: 302-047-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR