2'-Fluor-5'-methylacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Fluoro-5-methylphenyl)ethanone

006272-5G
FLUO006272-5GEA 0 EUR
FLUO006272-5G
2'-Fluor-5'-methylacetophenon
2'-Fluor-5'-methylacetophenon
Formel: C₉H₉FO
Molecular Weight: 152,17 g/mol
MDL: MFCD00236680
CAS-Nummer: 446-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR