5-Fluor-2-methylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-methylbenzolsulfonsäurechlorid, 5-Fluor-2-methylbenzol-1-sulfonylchlorid, 5-Fluor-2-methylbenzolsulfochlorid

006277-5G
FLUO006277-5GEA 42.9 EUR
FLUO006277-5G
5-Fluor-2-methylbenzolsulfonylchlorid
5-Fluor-2-methylbenzolsulfonylchlorid
Formel: C₇H₆ClFO₂S
Molecular Weight: 208,64 g/mol
Siedepunkt: 155…158 °C (50 mmHg)
MDL: MFCD00052958
CAS-Nummer: 445-05-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR