2-Fluor-3-methylbenzylbromid
Lieferant: FLUOROCHEM
Synonyme:
1-(Bromomethyl)-2-fluoro-3-methylbenzene
006281-1G
006281.5G
FLUO006281-1GEA
45.9
EUR
FLUO006281-1G
FLUO006281.5G
2-Fluor-3-methylbenzylbromid
2-Fluor-3-methylbenzylbromid
Formel:
C₈H₈BrF Siedepunkt: 105…106 °C (10 mmHg) |
MDL:
MFCD00042470 CAS-Nummer: 151412-12-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...