1-Fluoro-2-(2-nitrovinyl)benzene

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-beta-nitrostyrene, 1-Fluoro-2-(2-nitroethenyl)benzene

006305-5G
FLUO006305-5GEA 2250 EUR
FLUO006305-5G
1-Fluoro-2-(2-nitrovinyl)benzene
1-Fluoro-2-(2-nitrovinyl)benzene
Formel: C₈H₆FNO₂
Molecular Weight: 167,14 g/mol
MDL: MFCD00042451
CAS-Nummer: 399-25-7
EINECS: 206-915-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR