(4-Fluorphenoxy)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Fluorophenoxyacetic Acid

006318.5G 006318-25G
FLUO006318.5GEA 19.8 EUR
FLUO006318.5G FLUO006318-25G
(4-Fluorphenoxy)essigsäure
(4-Fluorphenoxy)essigsäure
Formel: (C₆H₄F)OCH₂COOH
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 106 °C
Dichte: 1,516 g/cm³ (20 °C)
MDL: MFCD00004304
CAS-Nummer: 405-79-8
EINECS: 206-974-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR