3-Fluor-α-methylbenzylalkohol
Lieferant: FLUOROCHEM
006338-5G
FLUO006338-5GEA
237
EUR
FLUO006338-5G
3-Fluor-α-methylbenzylalkohol
3-Fluor-α-methylbenzylalkohol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol |
MDL:
MFCD00014402 CAS-Nummer: 402-63-1 EINECS: 206-950-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...