3-(4-Fluorphenyl)propionsäure
Lieferant: FLUOROCHEM
006340-5G
FLUO006340-5GEA
84.9
EUR
FLUO006340-5G
3-(4-Fluorphenyl)propionsäure
3-(4-Fluorphenyl)propionsäure
Formel:
C₉H₉FO₂ |
MDL:
MFCD00060327 CAS-Nummer: 459-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...