3'-Fluorpropiophenon

Lieferant: FLUOROCHEM

Synonyme: m-Fluorpropiophenon

006359-5G 006359.10G
FLUO006359-5GEA 82.1 EUR
FLUO006359-5G FLUO006359.10G
3'-Fluorpropiophenon
3'-Fluorpropiophenon
Formel: C₉H₉FO
Molecular Weight: 152,17 g/mol
MDL: MFCD00061245
CAS-Nummer: 455-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR