2'-Fluor-3'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone

006373-1G 006373-5G
FLUO006373-1GEA 40.1 EUR
FLUO006373-1G FLUO006373-5G
2'-Fluor-3'-(trifluormethyl)acetophenon
2'-Fluor-3'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206.14 g/mol
MDL: MFCD00061145
CAS-Nummer: 207853-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR