2'-Fluor-5'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

006375-1G 006375-5G 006375-10G
FLUO006375-1GEA 51.8 EUR
FLUO006375-1G FLUO006375-5G FLUO006375-10G
2'-Fluor-5'-(trifluormethyl)acetophenon
2'-Fluor-5'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206.14 g/mol
MDL: MFCD00061248
CAS-Nummer: 202664-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR