α,α,α,2-Tetrafluor-m-toluylsäure

Lieferant: FLUOROCHEM

CFF631.5 CFF631.10 006394-1G 006394-5G 006394-25G
CHMPCFF631.5EA 268 EUR
CHMPCFF631.5 CHMPCFF631.10 FLUO006394-1G FLUO006394-5G FLUO006394-25G
α,α,α,2-Tetrafluor-m-toluylsäure
α,α,α,2-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
MDL: MFCD00040980
CAS-Nummer: 115029-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR