2-Fluor-4-(trifluormethyl)benzylalkohol

Lieferant: FLUOROCHEM

006410-1G 006410-5G 006410-10G
FLUO006410-1GEA 48.8 EUR
FLUO006410-1G FLUO006410-5G FLUO006410-10G
2-Fluor-4-(trifluormethyl)benzylalkohol
2-Fluor-4-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
MDL: MFCD00070808
CAS-Nummer: 197239-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR