4-Fluor-2-(trifluormethyl)benzylalkohol

Lieferant: FLUOROCHEM

006414-1G 006414-5G 006414-10G
FLUO006414-1GEA 30.5 EUR
FLUO006414-1G FLUO006414-5G FLUO006414-10G
4-Fluor-2-(trifluormethyl)benzylalkohol
4-Fluor-2-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
MDL: MFCD00061170
CAS-Nummer: 220227-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR