2-Fluor-5-(trifluormethyl)benzylbromid
Lieferant: FLUOROCHEM
006424-1G
006424-5G
FLUO006424-1GEA
36.9
EUR
FLUO006424-1G
FLUO006424-5G
2-Fluor-5-(trifluormethyl)benzylbromid
2-Fluor-5-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 22 °C |
MDL:
MFCD00061174 CAS-Nummer: 220239-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...