4-Fluor-2-(trifluormethyl)benzylbromid
Lieferant: FLUOROCHEM
006427-1G
006427-5G
006427-25G
FLUO006427-1GEA
34.1
EUR
FLUO006427-1G
FLUO006427-5G
FLUO006427-25G
4-Fluor-2-(trifluormethyl)benzylbromid
4-Fluor-2-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 22 °C |
MDL:
MFCD00061176 CAS-Nummer: 206860-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...