3-Fluor-5-(trifluormethyl)phenylacetonitril

Lieferant: FLUOROCHEM

006445-5G 006445-10G 006445-1G
FLUO006445-5GEA 237 EUR
FLUO006445-5G FLUO006445-10G FLUO006445-1G
3-Fluor-5-(trifluormethyl)phenylacetonitril
3-Fluor-5-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
MDL: MFCD00061181
CAS-Nummer: 239087-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR