5-Fluoruridin

Lieferant: FLUOROCHEM

Synonyme: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione

006461-100MG
FLUO006461-100MGEA 0 EUR
FLUO006461-100MG
5-Fluoruridin
5-Fluoruridin
Formel: C₉H₁₁FN₂O₆
Molecular Weight: 262,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036832
CAS-Nummer: 316-46-1
EINECS: 206-260-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR