Hexafluoraceton Trihydrat

Lieferant: FLUOROCHEM

006552-25G 006552-100G
FLUO006552-25GEA 60.9 EUR
FLUO006552-25G FLUO006552-100G
Hexafluoraceton Trihydrat
Hexafluoraceton Trihydrat
Formel: C₃F₆O·3H₂O
MDL: MFCD00149061
CAS-Nummer: 34202-69-2
EINECS: 211-676-3
UN: 2552
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR