2,2,3,3-Tetrachlorhexafluorbutan

Lieferant: FLUOROCHEM
Achtung

006576-5G
FLUO006576-5GEA 224 EUR
FLUO006576-5G
2,2,3,3-Tetrachlorhexafluorbutan
2,2,3,3-Tetrachlorhexafluorbutan
Formel: C₄Cl₄F₆
MDL: MFCD00042088
CAS-Nummer: 375-34-8
EINECS: 206-787-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR