Quecksilber(II)trifluormethansulfonat
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
Mercury(II) trifluoromethanesulphonate, Mercury triflate
006730-100G
FLUO006730-100GEA
0
EUR
FLUO006730-100G
Quecksilber(II)trifluormethansulfonat
Quecksilber(II)trifluormethansulfonat
Formel:
C₂F₆HgO₆S₂ Molecular Weight: 498,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144746 CAS-Nummer: 49540-00-3 UN: 2025 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...