Methyl-2,2-difluor-2-(fluorsulfonyl)acetat

Lieferant: FLUOROCHEM

006805-5G 006805-25G
FLUO006805-5GEA 55.4 EUR
FLUO006805-5G FLUO006805-25G
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel: C₃H₃F₃O₄S
MDL: MFCD00144316
CAS-Nummer: 680-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR