4'-Nitro-3'-(trifluormethyl)acetanilid
Lieferant: FLUOROCHEM
Synonyme:
N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide
006902-5G
FLUO006902-5GEA
84.5
EUR
FLUO006902-5G
4'-Nitro-3'-(trifluormethyl)acetanilid
4'-Nitro-3'-(trifluormethyl)acetanilid
Formel:
C₉H₇F₃N₂O₃ |
MDL:
MFCD00017994 CAS-Nummer: 393-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...