Palladium(II)trifluoracetat

Lieferant: FLUOROCHEM

Synonyme: Trifluoressigsäure Palladium(II)salz

006935-1G
FLUO006935-1GEA 218 EUR
FLUO006935-1G
Palladium(II)trifluoracetat
Palladium(II)trifluoracetat
Formel: C₄F₆O₄Pd
Molecular Weight: 332,45 g/mol
MDL: MFCD00013204
CAS-Nummer: 42196-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR