Perfluor-1-butansulfonsäurefluorid

Lieferant: FLUOROCHEM

007001-25G 007001-100G 007001-250G
FLUO007001-25GEA 36.9 EUR
FLUO007001-25G FLUO007001-100G FLUO007001-250G
Perfluor-1-butansulfonsäurefluorid
Perfluor-1-butansulfonsäurefluorid
Formel: C₄F₁₀O₂S
MDL: MFCD00007422
CAS-Nummer: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR