Perfluoro-1,2-dimethylcyclobutan
Lieferant: FLUOROCHEM
Synonyme:
1,2-Di(trifluormethyl)hexafluorcyclobutan, 1,1,2,2,3,4-Hexafluoro-3,4-di(trifluoromethyl)cyclobutane, Perfluor-1,2-dimethylcyclobutan, 1,2-Dimethylperfluorcyclobutan
007031-25G
FLUO007031-25GEA
233
EUR
FLUO007031-25G
Perfluoro-1,2-dimethylcyclobutan
Perfluoro-1,2-dimethylcyclobutan
Formel:
C₆F₁₂ Molecular Weight: 300,05 g/mol |
MDL:
MFCD00013737 CAS-Nummer: 2994-71-0 EINECS: 221-065-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...