1H,1H,2H,2H-Perfluor-1-iodhexan

Lieferant: FLUOROCHEM
Achtung

007066-25G 007066-100G 007066-5G
FLUO007066-25GEA 210 EUR
FLUO007066-25G FLUO007066-100G FLUO007066-5G
1H,1H,2H,2H-Perfluor-1-iodhexan
1H,1H,2H,2H-Perfluor-1-iodhexan
Formel: C₆H₄F₉I
Lagertemperatur: Kühlschrank
MDL: MFCD00039409
CAS-Nummer: 2043-55-2
EINECS: 218-055-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR