1H,1H,2H,2H-Tridecafluor-1-octanol

Lieferant: FLUOROCHEM

007128-25G 007128-100G
FLUO007128-25GEA 97.2 EUR
FLUO007128-25G FLUO007128-100G
1H,1H,2H,2H-Tridecafluor-1-octanol
1H,1H,2H,2H-Tridecafluor-1-octanol
Formel: C₈H₅F₁₃O
MDL: MFCD00042143
CAS-Nummer: 647-42-7
EINECS: 211-477-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR