Praseodymtrifluorid

Lieferant: FLUOROCHEM

007228-10G 007228-50G 007228-100G
FLUO007228-10GEA 0 EUR
FLUO007228-10G FLUO007228-50G FLUO007228-100G
Praseodymtrifluorid
Praseodymtrifluorid
Formel: F₃Pr
Molecular Weight: 197.9 g/mol
Schmelzpunkt: 1395
Dichte: 6.3 g/cm³ (20 °C)
MDL: MFCD00016287
CAS-Nummer: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR