2-(1,1,2,2-Tetrafluorethoxy)toluol

Lieferant: FLUOROCHEM

007348-10G
FLUO007348-10GEA 0 EUR
FLUO007348-10G
2-(1,1,2,2-Tetrafluorethoxy)toluol
2-(1,1,2,2-Tetrafluorethoxy)toluol
Formel: C₉H₈F₄O
MDL: MFCD00042103
CAS-Nummer: 42145-66-4
EINECS: 255-682-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR