4-(1,1,2,2-Tetrafluorethoxy)toluol

Lieferant: FLUOROCHEM

007350-10G
FLUO007350-10GEA 0 EUR
FLUO007350-10G
4-(1,1,2,2-Tetrafluorethoxy)toluol
4-(1,1,2,2-Tetrafluorethoxy)toluol
Formel: C₉H₈F₄O
Molecular Weight: 208,1528 g/mol
MDL: MFCD00042105
CAS-Nummer: 1737-11-7
EINECS: 217-086-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR