Thulium(III)fluorid
Lieferant: FLUOROCHEM
007426-1G
FLUO007426-1GEA
0
EUR
FLUO007426-1G
Thulium(III)fluorid
Thulium(III)fluorid
Formel:
F₃Tm Siedepunkt: ∼2200 °C (1013 hPa) Schmelzpunkt: 1158 °C Dichte: 7,9 g/cm³ (20 °C) |
MDL:
MFCD00049611 CAS-Nummer: 13760-79-7 EINECS: 237-353-7 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...