1,2,2-Tribrom-3,3,3-trifluorpropan

Lieferant: FLUOROCHEM

007437-25G
FLUO007437-25GEA 0 EUR
FLUO007437-25G
1,2,2-Tribrom-3,3,3-trifluorpropan
1,2,2-Tribromo-3,3,3-trifluoropropane
Formel: C₃H₂Br₃F₃
CAS-Nummer: 421-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR