2,4,5-Trifluorbenzylbromid
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
α-Brom-2,4,5-trifluortoluol
007505-5G
FLUO007505-5GEA
51.8
EUR
FLUO007505-5G
2,4,5-Trifluorbenzylbromid
2,4,5-Trifluorbenzylbromid
Formel:
C₇H₄BrF₃ Molecular Weight: 225,01 g/mol Siedepunkt: 182 °C (1013 hPa) Schmelzpunkt: 21 °C |
MDL:
MFCD00061209 CAS-Nummer: 157911-56-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...