4-(Trifluormethoxy)anisol
Lieferant: FLUOROCHEM
007532-1G
007532-5G
007532-25G
FLUO007532-1GEA
31.1
EUR
FLUO007532-1G
FLUO007532-5G
FLUO007532-25G
4-(Trifluormethoxy)anisol
4-(Trifluormethoxy)anisol
Formel:
C₈H₇F₃O₂ Siedepunkt: 163…165 °C (1013 hPa) Dichte: 1,272 g/cm³ (20 °C) |
MDL:
MFCD00216942 CAS-Nummer: 710-18-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...