2-(Trifluormethoxy)benzaldehyd

Lieferant: FLUOROCHEM
Achtung

007533-1G 007533-5G 007533-25G
FLUO007533-1GEA 31.1 EUR
FLUO007533-1G FLUO007533-5G FLUO007533-25G
2-(Trifluormethoxy)benzaldehyd
2-(Trifluormethoxy)benzaldehyd
Formel: C₈H₅F₃O₂
MDL: MFCD00042405
CAS-Nummer: 94651-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR