2-(Trifluormethoxy)benzamid

Lieferant: FLUOROCHEM

007536-5G 007536-25G
FLUO007536-5GEA 128 EUR
FLUO007536-5G FLUO007536-25G
2-(Trifluormethoxy)benzamid
2-(Trifluormethoxy)benzamid
Formel: C₈H₆F₃NO₂
MDL: MFCD00042407
CAS-Nummer: 127979-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR