2-(Trifluormethoxy)benzylalkohol

Lieferant: FLUOROCHEM

007549-1G 007549-5G
FLUO007549-1GEA 31.1 EUR
FLUO007549-1G FLUO007549-5G
2-(Trifluormethoxy)benzylalkohol
2-(Trifluormethoxy)benzylalkohol
Formel: C₈H₇F₃O₂
MDL: MFCD00153285
CAS-Nummer: 175278-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR