1-Brom-2-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

007555-25G 007555-10G 007555-50G
FLUO007555-25GEA 48.8 EUR
FLUO007555-25G FLUO007555-10G FLUO007555-50G
1-Brom-2-(trifluormethoxy)benzol
1-Brom-2-(trifluormethoxy)benzol
Formel: C₇H₄BrF₃O
MDL: MFCD00040942
CAS-Nummer: 64115-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR