1-Chlor-4-(trifluormethoxy)benzol
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)chlorobenzene
007556-5G
007556-10G
FLUO007556-5GEA
0
EUR
FLUO007556-5G
FLUO007556-10G
1-Chlor-4-(trifluormethoxy)benzol
1-Chloro-4-(trifluoromethoxy)benzene
Formel:
C₇H₄ClF₃O |
MDL:
MFCD00040953 CAS-Nummer: 461-81-4 EINECS: 207-315-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...