1-Chlor-4-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluoromethoxy)chlorobenzene

007556-5G 007556-10G
FLUO007556-5GEA 0 EUR
FLUO007556-5G FLUO007556-10G
1-Chlor-4-(trifluormethoxy)benzol
1-Chloro-4-(trifluoromethoxy)benzene
Formel: C₇H₄ClF₃O
MDL: MFCD00040953
CAS-Nummer: 461-81-4
EINECS: 207-315-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR