1-Nitro-3-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

007560-1G 007560-5G
FLUO007560-1GEA 0 EUR
FLUO007560-1G FLUO007560-5G
1-Nitro-3-(trifluormethoxy)benzol
1-Nitro-3-(trifluormethoxy)benzol
Formel: C₇H₄F₃NO₃
MDL: MFCD00041011
CAS-Nummer: 2995-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR