2'-(Trifluormethyl)acetanilid
Lieferant: FLUOROCHEM
Synonyme:
2-Acetamidobenzotrifluorid
007577-25G
FLUO007577-25GEA
107
EUR
FLUO007577-25G
2'-(Trifluormethyl)acetanilid
2'-(Trifluormethyl)acetanilid
Formel:
C₉H₈F₃NO |
MDL:
MFCD00013555 CAS-Nummer: 344-62-7 EINECS: 206-453-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...