4-(Trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluormethyl)benzolsulfonsäurechlorid, 4-(Trifluormethyl)benzol-1-sulfonylchlorid, 4-(Trifluormethyl)benzolsulfochlorid
007589-5G
007589-25G
007589-100G
FLUO007589-5GEA
36.9
EUR
FLUO007589-5G
FLUO007589-25G
FLUO007589-100G
4-(Trifluormethyl)benzolsulfonylchlorid
4-(Trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₄ClF₃O₂S Siedepunkt: 49,5…50.5 °C (0.2 mmHg) Schmelzpunkt: 31…33 °C |
MDL:
MFCD00042422 CAS-Nummer: 2991-42-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...