4-(Trifluormethyl)nicotinsäure

Lieferant: FLUOROCHEM

007629-1G 007629-5G
FLUO007629-1GEA 30.3 EUR
FLUO007629-1G FLUO007629-5G
4-(Trifluormethyl)nicotinsäure
4-(Trifluormethyl)nicotinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191.11 g/mol
MDL: MFCD00082626
CAS-Nummer: 158063-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR