2-(Trifluormethyl)phenylacetonitril
Lieferant: FLUOROCHEM
007635-5G
007635-100G
007635-25G
FLUO007635-5GEA
33.8
EUR
FLUO007635-5G
FLUO007635-100G
FLUO007635-25G
2-(Trifluormethyl)phenylacetonitril
2-(Trifluormethyl)phenylacetonitril
Formel:
C₉H₆F₃N Siedepunkt: 113…116 °C (23 mmHg) Schmelzpunkt: 29…30 °C |
MDL:
MFCD00001903 CAS-Nummer: 3038-47-9 EINECS: 221-239-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...