4-(Trifluormethyl)phenylacetonitril
Lieferant: FLUOROCHEM
007637-5G
007637-25G
007637-100G
FLUO007637-5GEA
0
EUR
FLUO007637-5G
FLUO007637-25G
FLUO007637-100G
4-(Trifluormethyl)phenylacetonitril
4-(Trifluormethyl)phenylacetonitril
Formel:
C₉H₆F₃N Siedepunkt: 131…132 °C (20 mmHg) Dichte: 1,535 g/cm³ (–150.15 °C) |
MDL:
MFCD00001921 CAS-Nummer: 2338-75-2 EINECS: 219-052-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...