1-(3-(Trifluormethyl)phenyl)imidazol

Lieferant: FLUOROCHEM

007649-1G 007649-5G
FLUO007649-1GEA 137 EUR
FLUO007649-1G FLUO007649-5G
1-(3-(Trifluormethyl)phenyl)imidazol
1-(3-(Trifluoromethyl)phenyl)imidazole
Formel: C₁₀H₇F₃N₂
MDL: MFCD00060487
CAS-Nummer: 25371-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR