3-(Trifluormethyl)pyridin

Lieferant: FLUOROCHEM

007663-1G 007663-5G 007663-10G
FLUO007663-1GEA 31.1 EUR
FLUO007663-1G FLUO007663-5G FLUO007663-10G
3-(Trifluormethyl)pyridin
3-(Trifluoromethyl)pyridine
Formel: C₆H₄F₃N
Molecular Weight: 147.1 g/mol
Siedepunkt: 110…113 °C (1013 hPa)
MDL: MFCD00100032
CAS-Nummer: 3796-23-4
EINECS: 223-268-2
UN: 1992
ADR: 3 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR